Learn More
Specac™ Heating Jacket/Controller for Cyclone™ C5
Product Code. 15230328
Encompass_Preferred
Click to view available options
Type:
Heating Jacket/Controller for Cyclone C5
Product Code. 15230328
Brand: Specac™ GS24305
This item requires a price update prior to being ordered.
Request a Price for this item here.
This heated jacket is compatible for the Cyclone C5 Long Pathlength Gas Cells. The jacket simply slides over the gas cell and can be operated from ambient temperatures up to 200°C.
Specifications
Type Heating Jacket/Controller for Cyclone C5