Learn More
Specac™ Heating Jacket/Controller for Cyclone™ C10
Description
This heated jacket is compatible for the Cyclone C10 long pathlength gas cells. The jacket simply slides over the gas cell and can be operated from ambient temperatures up to 200°C.
Specifications
Specifications
| Type | Heating Jacket/Controller for Cyclone C10 |