missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fistreem International™ Heating Element, Calypso Ultrapure Water Still
Description
- A high temperature cut-off protects the heaters in the event of water failure while operator safety is assured through totally enclosed glassware.
- The Calypso Still is economical and available with 2L/hr or 4L/hr outputs.
Specifications
Specifications
| For Use With (Equipment) | 2 Liter Model |
| Type | Heating Element |
| For Use With (Application) | Calypso Ultrapure Water Still |
| Voltage | 220 V |
| Wattage | 0.9 kw |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?