Learn More
Grant Instruments™ Heating Block for TS-DW Deep-well Microplate Thermoshaker
Description
Block for one deep-well plate
- Shaped to fit the microplates for better heat transfer
- Three year warranty
Specifications
Specifications
| For Use With (Equipment) | TS-DW Microplate Thermoshaker |
| Holds | Starlab 96 Deep-well Plate |
| Type | Block |
| Size | 1200 μL |