missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Heating Block

Product Code. 17862617 Shop All Corning Products
Change view
Click to view available options
Tube Capacity:
0.2 mL
0.5 mL
1.5 mL
2 mL
5 mL
15 mL
50 mL
Well Count:
24
4
8
96
96/384
Style:
Cone Bottom
Flat Bottom
Round Bottom
Unit Size:
Each
Product Code. Tube Capacity Well Count Style unitSize
17862617 0.2 mL 96 Flat Bottom Each
17872607 0.5 mL 24 Flat Bottom Each
17832617 1.5 mL 24 Cone Bottom Each
17882607 2 mL 24 Round Bottom Each
17852617 5 mL 8 Flat Bottom Each
17862607 5 mL 8 Cone Bottom Each
17842617 15 mL 8 Flat Bottom Each
17892607 50 mL 4 Flat Bottom Each
17802617 0.2 mL 96/384 Flat Bottom Each
9 options
This item is not returnable. View return policy

Product Code. 17862617

Brand: Axygen™ I4010H

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Interchangeable aluminum block to accommodate PCR plates and a variety of common life science vessels.

  • No additional tools are required for block exchanges
  • The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
  • The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
TRUSTED_SUSTAINABILITY

Specifications

Tube Capacity 0.2 mL
Weight (Metric) 0.3 kg
Style Flat Bottom
Well Count 96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.