missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Heating Block
Shop All Corning Products
Click to view available options
Tube Capacity:
0.2 mL
0.5 mL
1.5 mL
2 mL
5 mL
15 mL
50 mL
Well Count:
24
4
8
96
96/384
Style:
Cone Bottom
Flat Bottom
Round Bottom
Unit Size:
Each
Description
- No additional tools are required for block exchanges
- The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
- The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
- Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Specifications
Specifications
| Tube Capacity | 1.5 mL |
| Weight (Metric) | 0.3 kg |
| Style | Cone Bottom |
| Well Count | 24 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction