missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- No additional tools are required for block exchanges
- The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
- The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
- Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Specifications
Specifications
| Tube Capacity | 5 mL |
| Weight (Metric) | 0.3 kg |
| Style | Cone Bottom |
| Well Count | 8 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?