missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethyl(3,3,3-trifluoropropyl)silane, 97%

Product Code. 11344026
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11344026

Brand: Thermo Scientific Alfa Aesar L17292.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 358-67-8 | C6H13F3O2Si | 202.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 358-67-8
Molecular Formula C6H13F3O2Si
Molecular Weight (g/mol) 202.248
MDL Number MFCD00053175
InChI Key DIJRHOZMLZRNLM-UHFFFAOYSA-N
Synonym dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl
PubChem CID 67751
IUPAC Name dimethoxy-methyl-(3,3,3-trifluoropropyl)silane
SMILES CO[Si](C)(CCC(F)(F)F)OC

Specifications

Boiling Point 85°C (150mmHg)
Quantity 5 g
UN Number UN1993
Beilstein 1753237
Flash Point 58°C (136°F)
Formula Weight 202.25
Percent Purity 97%
Refractive Index 1.3576
Sensitivity Moisture sensitive
Density 1.1
Chemical Name or Material Dimethoxymethyl(3,3,3-trifluoropropyl)silane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.