Learn More
Biochrom Heated Cell Holder for Novaspec III+ and Novaspec Pro Spectrophotometers
Heated Cell Holder for Novaspec III+ and Novaspec Pro Spectrophotometers, for thermostatted measurements at 37°C.
Brand: Biochrom 80-3007-13
Additional Details : Weight : 0.20000kg
Description
- Heated cell holder
- For use with Biochrom Novaspec III+ and Novaspec Pro Spectrophotometers
Specifications
Novaspec III+ and Novaspec Pro Spectrophotometers | |
Reusable | |
Novaspec III+ and Novaspec Pro Spectrophotometers |
Heated Cell Holder | |
For Thermostatted Measurements at 37°C | |
37°C |