missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heat-Shrink Bands for Nalgene™ PETG Media Bottles

Product Code. 10602064
Change view
Click to view available options
Neck Finish:
20-415
24-415
38-430
53B
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Unit Size
10602064 24-415 Pack of 1000
10424553 20-415 Pack of 1000
10314533 38-430 Pack of 1000
10521465 53B Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10602064 Supplier Thermo Scientific™ Supplier No. 3121600240
  • ONLINE EXCLUSIVE PRICE
    £125.88
    £75.80 / Pack of 1000
    Save £50.08
    39% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure the integrity of bottle contents with these tamper-resistant seals. Heat-shrink bands are gamma stable and include a perforated tear strip feature for easy removal.

Ensure the integrity of bottle contents with Thermo Scientific™ Heat-Shrink Bands for Thermo Scientific™ Nalgene™ PETG Media Bottles. Simply apply the recommended application torque for Thermo Scientific Nalgene closures and slide the heat-shrink band over the closure and bottle neck. Heat the band to shrink and secure contents with a tamper-resistant seal.

  • Gamma stable bands include a perforated 'tear strip' feature for easy removal.
  • Bands available are in four sizes to mate with Nalgene PETG Square Media Bottles.
  • Bands are packed 1,000 per case (two zipper bags of 500 each).

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material PETG
No. per Case 1000
No. per Pack 1000
Sterility Non-sterile
Torque 12-17 in.-lb
Torque Wrench Fittings 2195-1024
Neck Finish 24-415
For Use With (Equipment) 60 mL Square Media Bottle
Autoclavable Not Autoclavable
Disposable Single-use
Product Type Heat-shrink Band
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.