Learn More
Heidolph™ Heat-On Multi-Well Holder Package
Product Code. 11761719 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Round Bottom Flasks
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11761719 Round Bottom Flasks Each
1 options
Product Code. 11761719 Supplier Heidolph™ Supplier No. 5058120000
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This package includes the Multi-Well Holder and 6 inserts for small round bottom flasks.
  • Heat-On inserts are suitable for two 25 mL, 50 mL, or 100 mL round bottom flasks
  • Multi-Well holder can enclose two Heat-On inserts at the same time, allowing for multiple combinations
  • In order to measure precisely with a temperature sensor, a small hole is provided on one side of the Heat-On Block
  • Scratch and chemical-resistant Multi-Well holder
  • Safety lifting handles are optional and available separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Round Bottom Flasks
For Use With (Application) Heating Glassware
Includes Holder and 6 Inserts
Temperature (Metric) 260°C
Coating PTFE
Material Aluminum