Learn More
Heidolph™ Heat-On Multi-Well Basic Package
Product Code. 11701719 Shop All Heidolph Instruments Products
Change view
Click to view available options
Type:
Heat-On Multi-Well Basic Package
Unit Size:
Each
Product Code. Type unitSize
11701719 Heat-On Multi-Well Basic Package Each
1 options
Product Code. 11701719
Brand: Heidolph™ 5058130000
Only null left
Add to basket
Add to basket
Heat-On attachment
  • Includes a multi-well holder and 3 inserts for small round bottom flasks
    • The Heat-On inserts are suitable for 25 mL, 50 mL and 100 mL round bottom flasks respectively
    • A max. of two flasks at a time can be received by the multi-well holder
  • The max. temperature for continuous use is 260°C
    • In order to measure this precisely with a temperature sensor, a small hole is provided on one side of the Heat-On Block
  • Safety lifting handles are optional and are available separately
Order Info
dimProductType
TRUSTED_SUSTAINABILITY
Specifications
Type Heat-On Multi-Well Basic Package
Coating Polymer
Material Aluminum