Learn More
Grant Instruments™ Heat Exchange Cooling Coil
Product Code. 11486487
Click to view available options
For Use With (Equipment):
OLS26
T100, TC120, TX150, TXF200
Unit Size:
Each
Product Code. 11486487
Brand: Grant Instruments™ CW5
Only null left
Add to basket
Add to basket
Can be attached to a cold water supply or refrigerated circulator. Grant Instruments™ Heat Exchange Coil maintains bath down to 2°C above the temperature of the coolant.
  • Stainless steel heat exchange coil that can be attached to cold water supply or a refrigerated circulator
  • Can be used to maintain a water bath temperature that is 2°C above the temperature of the coolant used
  • Compatible with OLS 26 Aqua Pro
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) T100, TC120, TX150, TXF200
Temperature Range 2°C
Type Heat Exchange Coil