missing translation for 'onlineSavingsMsg'
Learn More

Melford Heart Infusion Broth

Product Code. 30018323
Click to view available options
:
500 g
This item is not returnable. View return policy

Product Code. 30018323

Brand: Melford H11000500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended for the cultivations of rare microorganisms.

Recommended for the cultivations of rare microorganisms. Protect from moisture and light.

Brain Heart Infusion from solids: 2.0g

Casein Digest Peptone: 10.0g

Pepetic Digest of animal tissue: 5.0g

Yeast Extract: 3.0g

Sodium Chloride: 5.0g

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.