missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEAB Recombinant Rabbit Monoclonal Antibody (23GB3085), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA553597
This item is not returnable.
View return policy
Description
HEAB Recombinant Monoclonal antibody specifically detects HEAB in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| HEAB | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CLP1 | |
| AI462438; ATP/GTP-binding protein; cleavage and polyadenylation factor I subunit 1; CLP1; CLP1, cleavage and polyadenylation factor I subunit; CLP1, cleavage and polyadenylation factor I subunit, homolog; HAMAP-Rule:MF_03035}; hClp1; Heab; homolog of yeast CFIA subunit Clp1p; polyadenylation factor Clp1; polyadenylation factor Clp1 {ECO:0000255; Polynucleotide kinase Clp1; polynucleotide kinase Clp1 {ECO:0000255; polyribonucleotide 5'-hydroxyl-kinase Clp1; polyribonucleotide 5'-hydroxyl-kinase Clp1 {ECO:0000255; Pre-mRNA cleavage complex II protein Clp1; pre-mRNA cleavage complex II protein Clp1 {ECO:0000255; RGD1307679 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10978 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3085 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92989 | |
| CLP1 | |
| A synthesized peptide derived from HEAB (1-25AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction