missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEAB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104033
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CLP1.
HEAB is a protein coding gene.
Specifications
| HEAB | |
| Polyclonal | |
| Unconjugated | |
| CLP1 | |
| AI462438; ATP/GTP-binding protein; cleavage and polyadenylation factor I subunit 1; CLP1; CLP1, cleavage and polyadenylation factor I subunit; CLP1, cleavage and polyadenylation factor I subunit, homolog; HAMAP-Rule:MF_03035}; hClp1; Heab; homolog of yeast CFIA subunit Clp1p; polyadenylation factor Clp1; polyadenylation factor Clp1 {ECO:0000255; Polynucleotide kinase Clp1; polynucleotide kinase Clp1 {ECO:0000255; polyribonucleotide 5'-hydroxyl-kinase Clp1; polyribonucleotide 5'-hydroxyl-kinase Clp1 {ECO:0000255; Pre-mRNA cleavage complex II protein Clp1; pre-mRNA cleavage complex II protein Clp1 {ECO:0000255; RGD1307679 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10978, 311166, 98985 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5PQL4, Q92989, Q99LI9 | |
| CLP1 | |
| A synthesized peptide derived from human CLP1(Accession Q92989), corresponding to amino acid residues Y313-H363. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction