missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HE4/WFDC2 Antibody (404), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90132AF350
This item is not returnable.
View return policy
Description
HE4/WFDC2 Monoclonal specifically detects HE4/WFDC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HE4/WFDC2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Rat WFDC2 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 404 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human HE4/WFDC2 (NP_006094.3; Met1-Phe124). | |
| 0.1 mL | |
| Cancer | |
| 10406 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction