Learn More
HE4 Mouse anti-Human, CoraLite 488, Clone: 5B2D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66557-100UL
Description
The HE4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HE4. This antibody recognizes human antigen. The HE4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| HE4 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 10406 | |
| IgG1 |
| Immunofluorescence | |
| 5B2D9 | |
| Mouse | |
| Primary | |
| Human |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.