missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HE 99X Max Submarine Unit
Shop All Cytiva Products
Click to view available options
For Use With (Application):
Molecular biology
Unit Size:
Each
Description
The HE 99X Max Submarine Unit offers a variety of gel lengths and comb sizes to run exactly the gel you need.
- Cast gels 15cm wide x 10, 15, or 20cm long, with one or two combs
- Comb choices range from 1mL fragment purification sample, to 30-well comb for high-throughput PCR screening
- Adjustable comb back enables to control depth of sample wells
Unit Components:
- Seamless molded high impact plastic buffer tray
- Banana plugs safe from corrosion
- Guard to protect platinum electrodes
- Electrode carrier for simple electrode replacement
- Rugged molded lid with safety guards to protect against electrical hazard
- Large diameter bubble level and leveling feet
- Strain-relieved high voltage power cords
- Red and black details on running trays
- Buffer chamber for orientation
Specifications
Specifications
| For Use With (Application) | Molecular biology |
| Includes | Seamless molded high impact plastic buffer tray; Banana plugs; Guard to protect platinum electrodes; Rugged molded lid with safety guards to protect against electrical hazard; Large diameter bubble level and leveling feet; High voltage power cords |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction