Learn More
Masterflex™ HDPE T-Connector Barbed Fittings
Product Code. 11744769
Click to view available options
For Use With:
1/4 in. tubing I.D.
3/16 in. tubing I.D.
3/4 in. tubing I.D.
3/8 in. tubing I.D.
5/16 in. tubing I.D.
Unit Size:
Pack of 10
Product Code. 11744769
Brand: Masterflex™ 3062360
Only null left
Add to basket
Add to basket
Barbed T connectors are simple and economical fittings designed for quick and easy assembly of fluid paths in the laboratory.
  • Use with low-pressure applications, generally less than 30 psi (2 bar)
  • Manufactured from quality HDPE
  • Sterilization by ethylene oxide (ETO) only
  • Complete range of standard sizes available
  • Provided in packs of 10
Specifications
Material HDPE
For Use With 3/4 in. tubing I.D.
Packaging Quantity 10 Pack
Sterilization Method Ethylene Oxide
Shape T-shape
Product Type Barbed Fitting