missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ HDPE Straight Tubing Adapter

Product Code. 11828391 Shop All Brand Products
Click to view available options
Tubing Size:
11 to 14mm I.D.
18 to 20.5mm I.D
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11828391

Brand: BRAND™ 152510

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Constructed of HDPE. Straight tubing adapter includes hose barbs for a secure fit.

  • High-density Polyethylene construction
  • Straight tubing adapter includes a large and small end to accommodate many sizes.
  • Hose barbs provide a secure fit.
TRUSTED_SUSTAINABILITY

Specifications

Material HDPE
Overall Length (Metric) 73 mm
For Use With 12 to 14/18 to 20.5mm tubing I.D.
Packaging Quantity 10 Pack
Tubing Size 18 to 20.5mm I.D
Shape Straight
Product Type Tubing Adapter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.