Learn More
Masterflex™ Straight Connector Barbed Fitting, HDPE
Product Code. 11755559
Change view
Click to view available options
For Use With:
1/2 in. tubing I.D.
1/4 in. tubing I.D.
3/16 in. tubing I.D.
3/8 in. tubing I.D.
5/16 in. tubing I.D.
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11755559 1/2 in. tubing I.D. Pack of 10
11795549 1/4 in. tubing I.D. Pack of 10
11745559 3/8 in. tubing I.D. Pack of 10
11735559 5/16 in. tubing I.D. Pack of 10
11785549 3/16 in. tubing I.D. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11755559 Supplier Masterflex™ Supplier No. 3062192
Add to basket
Add to basket
Combine these simple, economical HDPE straight connector with flexible tubing for use in your low-pressure applications (generally less than 30 psi). HDPE Straight Connector Barbed Fittings slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
  • Designed for quick and easy assembly without tools
  • Very Good chemical resistance
  • Temp range: - 53 to 65°C
  • Sterilize by ethylene oxide
Specifications
Material HDPE
For Use With 1/2 in. tubing I.D.
Packaging Quantity 10 Pack
Product Type Barbed Fitting