Learn More
Masterflex™ Straight Connector Barbed Fitting, HDPE
Description
- Designed for quick and easy assembly without tools
- Very Good chemical resistance
- Temp range: - 53 to 65°C
- Sterilize by ethylene oxide
Specifications
Specifications
| Material | HDPE |
| For Use With | 5/16 in. tubing I.D. |
| Packaging Quantity | 10 Pack |
| Product Type | Barbed Fitting |