Learn More
Kartell SpA GL32 Screw Caps
Description
Standardized screw caps for polyethylene bottles.
Specifications
Specifications
| Color | Opaque White |
| Material | HDPE |
| Sterility | Non-sterile |
| Standard Taper Joint | GL32 |
| Disposable | Reusable |
| Fit for Purpose | Certified Quality |
| Product Type | Screw Cap |
| Autoclavable | Not Autoclavable |