missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE Screw Closure for Venting System

Product Code. 15156665
Click to view available options
:
Color: Black

Product Code. 15156665

Brand: Kautex™ 2000086365

Only null left
Add to basket
Add to basket

The HDPE Screw Closure for Venting System is ideal for use with jerrycans.

  • Accomodates venting system for jerrycans.
  • Black HDPE.
  • Tamper-evident ring.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance None
Closure Material HDPE
Closure Size 45 mm (DIN45)
Closure Type Screw Closure with Tamper-evident Ring and Venting System
Color Black
For Use With (Equipment) None
Material HDPE
Product Type Closure
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.