missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL HDPE Screw Cap
Description
- Standardized screw caps for PP bottles DIN 168
- Suitable for bottles with GL45 neck and Kartell's bottles codes 1612 and 1614.
Specifications
Specifications
| Color | White |
| Material | HDPE |
| Quantity | 1 pc. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL45 |
| Neck Finish | 61 mm |
| Certifications/Compliance | DIN 168 |
| Disposable | Reusable |
| Fit for Purpose | Certified Quality |
| Product Type | Screw Cap |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?