missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE 308/310 Series Narrow Neck Bottle

Product Code. 15500981
Click to view available options
Capacity (Metric):
0.5 L
1 L
2.5 L
Color:
Black
Natural
Unit Size:
Each
Pack of 120
Pack of 26
Pack of 64

Product Code. 15500981

Brand: Kautex™ 2000084152

Only null left
Add to basket
Add to basket

Use this Kautex™HDPE 308/310 Series Narrow Neck Bottle for packaging, sending samples, mailing and storage. Suitable for filling and transport of hazardous liquids, it features a recessed labeling field with sizes 1000 and 2500 mL. and is UN approved for liquids.

  • Optimized handle for safe and comfortable handling.
  • Recessed labelling field with sizes 1000 and 2500 mL.
  • GL 45 neck, three different types of closures available
  • leakproof
  • UN approved for liquids
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 L
Color Natural
Diameter (Metric) Inner Neck 28.7 mm
For Use With (Application) Packaging, Sending Samples, Mailing, Storage
Material HDPE
Neck Finish Narrow
Certifications/Compliance UN 3H1/X1.9/250
Mouth Narrow
Height (Metric) 211.8 mm
Product Type Square Narrow Mouth Bottle
Diameter (Metric) 96 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.