missing translation for 'onlineSavingsMsg'
Learn More

SCAT Collecting Tray, HDPE-EC

SCAT Collecting Trays store your waste containers and provide additional safety in case of any leakage.

Brand:  SCAT 117986

267.58 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10222480

  • £331.00 / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

  • Suitable for a wide range of canisters
  • Collect liquid in case of a leakage
  • Available with base insert so the canisters stay always clean and dry
  • Optional with electrostatic conductive material
  • Made of PE-HD or optional PE-HD-ec
Specifications

Specifications

24 L
Collecting tray with base insert, PE-HD electrostatic conductive, dimensions (W x H x D): 295 x 200 x 415 mm (interior), dimensions (W x H x D): 365 x 240 x 490 mm (exterior)
200 mm (Inner), 240 mm (Outer)
HDPE
295 (Inner) mm, 365 mm (Outer)
Handling of Hazardous Substances
1 Pc.
Rectangular
Inner: 415 x 295 x 200 mm - Outer: 490 x 365 x 240 mm
Black
415 (Inner) mm, 490 mm (Outer)
Collection Tray
Waste Containers
Base Insert
Electrically Conductive HDPE
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers