missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDLBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04382-20UL
This item is not returnable.
View return policy
Description
HDLBP Polyclonal specifically detects HDLBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HDLBP | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| HBPFLJ16432, HDL-binding protein, high density lipoprotein binding protein, High density lipoprotein-binding protein, PRO2900, VGLvigilin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1035-1268 of human HDLBP (NP_976221.1). | |
| 20 μL | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 3069 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction