missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDJ2 Polyclonal Antibody

Product Code. 16393945
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16393945 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16393945 Supplier Invitrogen™ Supplier No. PA587091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDJ2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJA1
Gene Accession No. P31689, P63036, P63037
Gene Alias DJ-2; DjA1; DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ heat shock protein family (Hsp40) member A1; dnaJ homolog subfamily A member 1; DnaJ protein homolog 2; Dnaj2; DNAJA1; dnaJ-like protein 1; DnaJ-like protein 2; DnaJ-like subfamily A member 1; h HSJ-2; HDJ2; hDj-2; Heat shock 40 kDa protein 4; heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ2; HSJ-2; HSPF4; human DnaJ protein 2; I79_019991; LOC100346508; LOW QUALITY PROTEIN: dnaJ homolog subfamily A member 1; NEDD7; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7; Rdj1
Gene Symbols DNAJA1
Host Species Rabbit
Immunogen Recombinant fusion protein of human DNAJA1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15502, 3301, 65028
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.