missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDHD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HDHD2 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16804685
|
Proteintech
26147-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16894675
|
Proteintech
26147-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDHD2 is a protein coding gene.Specifications
| HDHD2 | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q3UGR5, Q9H0R4 | |
| 76987, 84064 | |
| HDHD2 Fusion Protein Ag23518 | |
| Primary | |
| -20°C | |
| HDHD2 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 3110052N05Rik, HDHD2 | |
| Hdhd2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title