missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDGFL1 Monoclonal Antibody (249308)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561052
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| HDGFL1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| dJ309H15.1; HDGF (hepatoma-derived growth factor) like; HDGFL1; hepatoma derived growth factor-like 1; hepatoma-derived growth factor-like protein 1; PWWP domain containing 1; PWWP domain-containing protein 1; PWWP1; testicular tissue protein Li 84 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 15192 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 249308 | |
| Unconjugated | |
| HDGFL1 | |
| HDGFL1 | |
| E. coli-derived recombinant human HRP-1, Met1-Leu283. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction