Learn More
Invitrogen™ HDGF Recombinant Rabbit Monoclonal Antibody (JE56-32)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542522
Description
Positive Control: Rat cerebellum tissue lysate, SK-Br-3 cell lysate, A549 cell lysate, mouse cerebellum tissue lysate, F9, human liver carcinoma tissue, human cervix tissue, human colon carcinoma tissue, human breast carcinoma tissue, human esophagus tissue, human stomach carcinoma tissue, human small intestine tissue, THP-1. Subcellular Location: Nucleus, Cytoplasm, Extracellular exosome.
HDGF is a member of the hepatoma-derived growth factor family. This protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation.
Specifications
| HDGF | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P51858, P51859, Q8VHK7 | |
| HDGF | |
| Recombinant protein within human HDGF aa 50-200. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE56-32 | |
| Unconjugated | |
| HDGF | |
| AI118077; D3Ertd299e; DKFZp686J1764; FLJ96580; Hdgf; Hepatoma-derived growth factor; high mobility group protein 1-like 2; HMG1L2; HMG-1L2; Tdrm1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 114499, 15191, 3068 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.