missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDGF Antibody (PCRP-HDGF-1D1) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26942JF585
This item is not returnable.
View return policy
Description
HDGF Monoclonal antibody specifically detects HDGF in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
Specifications
| HDGF | |
| Monoclonal | |
| Janelia Fluor 585 | |
| FLJ96580, hepatoma-derived growth factor, hepatoma-derived growth factor (high-mobility group protein 1-like), High mobility group protein 1-like 2, HMG-1L2, HMG1L2DKFZp686J1764 | |
| Recombinant full-length human HDGF protein | |
| 0.1 mL | |
| Cancer, Cell Biology | |
| 3068 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-HDGF-1D1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction