missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDGF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71924
This item is not returnable.
View return policy
Description
HDGF Polyclonal specifically detects HDGF in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications
| HDGF | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| FLJ96580, hepatoma-derived growth factor, hepatoma-derived growth factor (high-mobility group protein 1-like), High mobility group protein 1-like 2, HMG-1L2, HMG1L2DKFZp686J1764 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3068 | |
| Store at 4C. Do not freeze. |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 2-10 mcg/mg, Immunohistochemistry-Paraffin 1:1000-1:5000, Western Blot (Negative) | |
| NP_004485.1 | |
| HDGF | |
| The immunogen for this product maps to a region between residue 100 and 150 of human Hepatoma-Derived Growth Factor using the numbering given in entry NP_004485.1 (GeneID 3068). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction