missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDDC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559342
This item is not returnable.
View return policy
Description
Immunogen sequence: PKLERKRLQV EQAPHSSPGA KLVKLADKLY NLRDLNRCTP EGWSEHRVQE YFEWAAQVVK GLQGTNRQLE EALKHLFKQR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
HDDC3 is involved in biological processes as a hydrolyzing enzyme in starvation response.
Specifications
| HDDC3 | |
| Polyclonal | |
| Unconjugated | |
| Hddc3 | |
| (ppGpp)ase; 1110033O09Rik; C86475; Guanosine-3',5'-bis(diphosphate) 3'-pyrophosphohydrolase MESH1; HD domain containing 3; HD domain-containing protein 3; HD domain-containing protein 3; guanosine-3',5'-bis(diphosphate) 3'-pyrophosphohydrolase MESH1; HDDC3; MESH1; Metazoan SpoT homolog 1; metazoan SpoT homolog-1; penta-phosphate guanosine-3'-pyrophosphohydrolase; RGD1311839 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 374659 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N4P3 | |
| Hddc3 | |
| Recombinant protein corresponding to Human HDDC3. Recombinant protein control fragment (Product #RP-98177). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction