missing translation for 'onlineSavingsMsg'
Learn More

HDDC3 Antibody, Novus Biologicals™

Product Code. 18339500 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339500 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339500 Supplier Novus Biologicals Supplier No. NBP287556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HDDC3 Polyclonal specifically detects HDDC3 in Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDDC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias (ppGpp)ase, EC 3.1.7.2, guanosine-3′-5′-bis(diphosphate) 3′-pyrophosphohydrolase MESH1, HD domain containing 3, HD domain-containing protein 3, MESH1, Metazoan SpoT homolog 1, metazoan SpoT homolog-1, MGC45386, Penta-phosphate guanosine-3′-pyrophosphohydrolase
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of Rat HDDC3
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 374659
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.