missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDDC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544917
This item is not returnable.
View return policy
Description
Peptide sequence: ASVSSATFSG HGARSLLQFL RLVGQLKRVP RTGWVYRNVQ RPESVSDHMY Sequence homology: Human: 100%.
Catalyzes the dephosphorylation of the nucleoside 5'-monophosphates deoxyadenosine monophosphate (dAMP), deoxycytidine monophosphate (dCMP), deoxyguanosine monophosphate (dGMP) and deoxythymidine monophosphate (dTMP). [UniProt].
Specifications
| HDDC2 | |
| Polyclonal | |
| Unconjugated | |
| Hddc2 | |
| 2310057G13Rik; C6orf74; CGI-130; dJ167O5.2; HCV NS5A-transactivated protein 2; HD domain containing 2; HD domain-containing protein 2; HDDC2; Hepatitis C virus NS5A-transactivated protein 2; NS5ATP2; RGD1307562; testicular tissue protein Li 83 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51020 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z4H3 | |
| Hddc2 | |
| Synthetic peptide directed towards the N-terminal of human HDDC2 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction