missing translation for 'onlineSavingsMsg'
Learn More

HDC Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18312874 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18312874 20 μL 20µL
18365986 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18312874 Supplier Novus Biologicals Supplier No. NBP30504820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HDC Polyclonal specifically detects HDC in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen HDC
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias histidine decarboxylase, MGC163399
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 453-662 of human HDC (NP_002103.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3067
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.