missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107049
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 85%; Dog: 92%; Guinea Pig: 79%; Horse: 92%; Human: 100%; Mouse: 92%; Rat: 92%.
Papillomavirus Regulatory Factor 1/HDBP2 is a novel transcription factor shuttling between nucleus and cytoplasm and binds to the specific GCCGGCG, which is an essential cis-element for Huntington's disease gene expression.
Specifications
| HDBP2 | |
| Polyclonal | |
| Unconjugated | |
| ZNF395 | |
| BC053701; Gm910; HD gene regulatory region-binding protein 2; HDBP2; HDBP-2; HD-regulating factor 2; HDRF-2; Huntington disease gene regulatory region-binding protein 2; Huntington's disease gene regulatory region-binding protein 2; papillomavirus regulatory factor; papillomavirus regulatory factor 1; papillomavirus regulatory factor PRF-1; papillomavirus-binding factor; PBF; PRF1; PRF-1; Si-1-8-14; Zfp395; Zinc finger protein 395; ZNF395 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9H8N7 | |
| ZNF395 | |
| Synthetic peptide directed towards the C-terminal region of human ZNF395. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction