missing translation for 'onlineSavingsMsg'
Learn More

HDACs/mTOR Inhibitor 1, MedChemExpress

Product Code. 16471087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471087

Brand: Medchem Express HY114414

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HDACs/mTOR Inhibitor 1 is a dual Histone Deacetylases (HDACs) and mammalian target of Rapamycin (mTOR) target inhibitor for treating hematologic malignancies, with IC50s of 0.19 nM, 1.8 nM, 1.2 nM and >500 nM for HDAC1, HDAC6, mTOR and PI3Kα, respectively. HDACs/mTOR Inhibitor 1 stimulates cell cycle arrest in G0/G1 phase and induce tumor cell apoptosis with low toxicity in vivo.

HDACs/mTOR Inhibitor 1 is a dual Histone Deacetylases (HDACs) and mammalian target of Rapamycin (mTOR) target inhibitor for treating hematologic malignancies, with IC50s of 0.19 nM, 1.8 nM, 1.2 nM and >500 nM for HDAC1, HDAC6, mTOR and PI3Kα, respectively. HDACs/mTOR Inhibitor 1 stimulates cell cycle arrest in G0/G1 phase and induce tumor cell apoptosis with low toxicity in vivo.

Specifications

Chemical Name or Material HDACs/mTOR Inhibitor 1
CAS 2271413-06-8
Color White
Purity Grade Notes Research
Molecular Formula C28H38N8O5
Quantity 50 mg
Solubility Information DMSO : 32.5 mg/mL (57.35 mM; Need ultrasonic)
SMILES O=C(NO)CCCCCCN1C2=NC(C3=CC=C(NC(N4C[C@@H](C)OCC4)=O)C=C3)=NC(N5CCOCC5)=C2C=N1
Molecular Weight (g/mol) 566.65
Formula Weight 566.65
Percent Purity 98.21%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.