missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC9 Antibody (OTI7G2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71493AF488
This item is not returnable.
View return policy
Description
HDAC9 Monoclonal specifically detects HDAC9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HDAC9 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HDAC9 | |
| Recombinant protein fragment corresponding to amino acids 181-460 of human HDAC9 (NP_055522) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7G2 | |
| Western Blot, Immunohistochemistry | |
| EC 3.5.1.98, HD7b, HD7HDAC, HD9, HDAC7, HDAC7BDKFZp779K1053, HDRP, histone deacetylase 4/5-related protein, Histone deacetylase 7B, histone deacetylase 9, Histone deacetylase-related protein, KIAA0744HDAC9B, MEF-2 interacting transcription repressor (MITR) protein, MEF2-interacting transcription repressor MITR, MITRHDAC9FL | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Epigenetics, Histone Deacetylases | |
| 9734.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction