missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511242
This item is not returnable.
View return policy
Description
HDAC7 Polyclonal Antibody for Western Blot
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| HDAC7 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8WUI4 | |
| HDAC7 | |
| KLH conjugated synthetic peptide between 920-952 amino acids from the C-terminal region of human HDAC7. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| HDAC7 | |
| 5830434K02Rik; HD7; HD7a; hdac4l; Hdac7; hdac-7; Hdac7a; histone deacetylase 7; Histone deacetylase 7A; mFLJ00062 | |
| Rabbit | |
| Size-exclusion - Dialysis, Ammonium sulfate precipitation | |
| RUO | |
| 51564 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction