missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16167-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to HDAC6. It does not cross react with other HDACs.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.Specifications
| HDAC6 | |
| Polyclonal | |
| Unconjugated | |
| HDAC6 | |
| HD6, HDAC6, histone deacetylase 6, JM21, KIAA0901 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10013, 15185, 84581 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UBN7, Q9Z2V5 | |
| Hdac6 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction