missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody (PCRP-HDAC6-1A4) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14206JF585
This item is not returnable.
View return policy
Description
HDAC6 Monoclonal antibody specifically detects HDAC6 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| HDAC6 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| Recombinant full-length human HDAC6 protein (Uniprot: Q9UBN7) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Neuroscience | |
| 10013 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunoprecipitation, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-HDAC6-1A4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction