missing translation for 'onlineSavingsMsg'
Learn More

HDAC6 Antibody (PCRP-HDAC6-1A4), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18341735 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341735 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341735 Supplier Novus Biologicals Supplier No. NBP314206JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HDAC6 Monoclonal antibody specifically detects HDAC6 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC6
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-HDAC6-1A4
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901
Host Species Mouse
Immunogen Recombinant full-length human HDAC6 protein (Uniprot: Q9UBN7)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10013
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.