missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £417.00
Specifications
| Antigen | HDAC6 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697338
|
Novus Biologicals
NBP2-41286-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18145068
|
Novus Biologicals
NBP2-41286 |
0.1 mg |
£417.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| HDAC6 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| HDAC6 | |
| IgG | |
| Affinity Purified | |
| 32 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 10013 | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human HDAC6. The immunogen is located within the last 50 amino acids of HDAC6. Amino Acid Squence: NIAHQNKFGEDMPHPH | |
| Primary | |
| Multiple isoforms of HDAC6 are known to exist. HDAC6 antibody is predicted to not cross-react with other members of the HDAC family. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title