missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16166-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody only binds HDAC5. It does not cross-react with other HDACs.
HDAC5 is a member of the class II mammalian histone deacetylase family, which is structurally related to yeast HDA1. Human HDAC5 is composed of 1122 amino acid residues. The deacetylase domain of HDAC5 is located at the C-terminal half of the molecule. The N-terminal non-deacetylase domain does not show any significant homology with any published sequence.Both domains are required for HDAC5-mediated repression of gene transcription. HDAC5 interacts with a growing number of transcriptional factors including MEF2A as well as other HDAC proteins. The interacting complexes bind to specific regions of chromatin and regulate gene transcription in these regions.Specifications
| HDAC5 | |
| Polyclonal | |
| Unconjugated | |
| HDAC5 | |
| Antigen NY CO 9, HD5, HDAC5, histone deacetylase 5, KIAA0600, NY CO 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10014, 15184, 84580 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UQL6, Q9Z2V6 | |
| Hdac5 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction