missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583172
This item is not returnable.
View return policy
Description
Immunogen sequence: THPEETEEEL TEQQEVLLGE GALTMPREGS TESESTQEDL EEEDEEDDGE EEEDCIQVKD EEGESGAEEG PDLEEPGAGY KKLFSDAQP Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 84%.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene belongs to the class II histone deacetylase/acuc/apha family. It possesses histone deacetylase activity and represses transcription when tethered to a promoter. It coimmunoprecipitates only with HDAC3 family member and might form multicomplex proteins. It also interacts with myocyte enhancer factor-2 (MEF2) proteins, resulting in repression of MEF2-dependent genes. This gene is thought to be associated with colon cancer. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| HDAC5 | |
| Polyclonal | |
| Unconjugated | |
| HDAC5 | |
| AI426555; Antigen NY-CO-9; DKFZp779K1053; FLJ90614; HD5; HD7; HD7b; HD9; HDAC; Hdac4; HDAC5; HDAC-5; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDRP; histone deacetylase 4; histone deacetylase 5; histone deacetylase mHDA1; KIAA0600; KIAA0744; mHDA1; mKIAA0600; NY-CO-9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UQL6 | |
| HDAC5 | |
| Recombinant protein corresponding to Human HDAC5. Recombinant protein control fragment (Product #RP-95274). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction