missing translation for 'onlineSavingsMsg'
Learn More

HDAC5 Antibody, Novus Biologicals™

Product Code. 18428600 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428600 25 μL 25µL
18449390 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18428600 Supplier Novus Biologicals Supplier No. NBP18343625ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HDAC5 Polyclonal antibody specifically detects HDAC5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC5
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias Antigen NY-CO-9, EC 3.5.1.98, FLJ90614, HD5, histone deacetylase 5, KIAA0600, NY-CO-9
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: THPEETEEELTEQQEVLLGEGALTMPREGSTESESTQEDLEEEDEEDDGEEEEDCIQVKDEEGESGAEEGPDLEEPGAGYKKLFSDAQP
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 10014
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.